missing translation for 'onlineSavingsMsg'
Learn More

2-Oxa-7-azaspiro[3.5]nonane hemioxalate, 97%

Catalog No. p-7052885
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57834MD 250 mg
AAH57834ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57834MD Supplier Thermo Scientific Chemicals Supplier No. H57834MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1429056-28-9 | C16H28N2O6 | 344.408 g/mol

Spiro forms of oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photo chromic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Spiro forms of oxazines find applications as leuco dyes, by frequently displaying chromism - reversibly interchanging between their colorless and colored forms. Spiro compounds are used as photo chromic materials.

Notes
Hygroscopic. Store at room temperature.

Chemical Identifiers

CAS 1429056-28-9
Molecular Formula C16H28N2O6
Molecular Weight (g/mol) 344.408
MDL Number MFCD11035888
InChI Key WWVUFRRXXSVWBJ-UHFFFAOYSA-N
Synonym 2-oxa-7-azaspiro 3.5 nonane oxalate 2:1, 2-oxa-7-azaspiro 3.5 nonane hemioxalate, bis 2-oxa-7-azaspiro 3.5 nonane ; oxalic acid, 2-oxa-7-azaspiro 3.5 nonan hemioxalate, 2-oxa-7-azaspiro 3,5 nonane hemioxalate, bis 2-oxa-7-azaspiro 3.5 nonane oxalate, 2-oxa-7-azaspiro 3.5 nonane hemi-oxalate
PubChem CID 71463619
IUPAC Name 2-oxa-7-azaspiro[3.5]nonane;oxalic acid
SMILES C1CNCCC12COC2.C1CNCCC12COC2.C(=O)(C(=O)O)O

Specifications

Melting Point 208°C
Color White to Gray
Density 1.048 g/mL
Quantity 250 mg
Formula Weight 172.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Oxa-7-azaspiro[3.5]nonane hemioxalate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only