missing translation for 'onlineSavingsMsg'
Learn More

2-Oxa-6-azaspiro[3.4]octane, 95%

Catalog No. p-7038581
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57579MD 250 mg
AAH57579ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57579MD Supplier Thermo Scientific Chemicals Supplier No. H57579MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 220290-68-6 | C6H11NO | 113.16 g/mol

2-Oxa-6-azaspiro[3.4]octane is involved in the preparation of azaspirocycle or azetidine substituted 4-anilinoquinazoline derivatives, which exhibits epidermal growth factor receptor (EGFR) inhibitory activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Oxa-6-azaspiro[3.4]octane is involved in the preparation of azaspirocycle or azetidine substituted 4-anilinoquinazoline derivatives, which exhibits epidermal growth factor receptor (EGFR) inhibitory activities.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids, reducing agents and bases.

Chemical Identifiers

CAS 220290-68-6
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD14586455
InChI Key ZHAIMJRKJKQNQI-UHFFFAOYSA-N
Synonym 2-oxa-6-azaspiro 3.4 octane, 2-oxa-6-aza-spiro 3.4 octane, 2-oxa-6-azaspiro 3,4 octane, acmc-209fqv, 2-oxa-7-azaspiro 3.4 octane, 2-oxa-6-aza-spiro 3,4 octane, spiro pyrrolidine-3,3'-oxetane
PubChem CID 53438323
IUPAC Name 2-oxa-7-azaspiro[3.4]octane
SMILES C1CNCC12COC2

Specifications

Density 1.08
Boiling Point 187°C
Flash Point 63°C (145°F)
Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 113.16
Percent Purity 95%
Chemical Name or Material 2-Oxa-6-azaspiro[3.4]octane
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only