Learn More
2-Oxa-6-azaspiro[3.3]heptane oxalate, 97%
CAS: 1159599-99-1 | C7H11NO5 | 189.17 g/mol
Supplier: Thermo Scientific Chemicals H6357803
Description
2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
| 2-Oxa-6-azaspiro[3.3]heptane oxalate | |
| C7H11NO5 | |
| 1 g | |
| Slightly soluble in water. | |
| OC(=O)C(O)=O.C1NCC11COC1 | |
| 189.17 | |
| 189.17 |
| 1159599-99-1 | |
| MFCD11976178 | |
| 2-oxa-6-azaspiro 3.3 heptane oxalate, 2-oxa-6-aza-spiro 3.3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3,3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3.3 heptane xoxalate, 2-oxa-6-azaspiro 3.3 heptane oxalic acid, 2-oxa-6-azaspiro 3.3 heptane; oxalic acid, 2-oxa-6-azaspiro 3.3 heptane ethanedioate, 2-oxa-6-azaspiro 3.3 heptane oxalate 1:1, 2-oxa-6-azaspiro 3.3 heptane, ethanedioate 1:1, acmc-2099rj | |
| KOUVDKDABFOPIG-UHFFFAOYSA-N | |
| 2-oxa-6-azaspiro[3.3]heptane;oxalic acid | |
| 53308464 | |
| 97% |
Chemical Identifiers
| 1159599-99-1 | |
| 189.17 | |
| KOUVDKDABFOPIG-UHFFFAOYSA-N | |
| 53308464 |
| C7H11NO5 | |
| MFCD11976178 | |
| 2-oxa-6-azaspiro 3.3 heptane oxalate, 2-oxa-6-aza-spiro 3.3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3,3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3.3 heptane xoxalate, 2-oxa-6-azaspiro 3.3 heptane oxalic acid, 2-oxa-6-azaspiro 3.3 heptane; oxalic acid, 2-oxa-6-azaspiro 3.3 heptane ethanedioate, 2-oxa-6-azaspiro 3.3 heptane oxalate 1:1, 2-oxa-6-azaspiro 3.3 heptane, ethanedioate 1:1, acmc-2099rj | |
| OC(=O)C(O)=O.C1NCC11COC1 |
Safety and Handling
GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only