missing translation for 'onlineSavingsMsg'
Learn More

2-Oxa-6-azaspiro[3.3]heptane oxalate, 97%

Catalog No. AAH6357806
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6357806 5 g
AAH63578MD 250 mg
AAH6357803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6357806 Supplier Thermo Scientific Chemicals Supplier No. H6357806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1159599-99-1 | C7H11NO5 | 189.17 g/mol

2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 1159599-99-1
Molecular Formula C7H11NO5
Molecular Weight (g/mol) 189.17
MDL Number MFCD11976178
InChI Key KOUVDKDABFOPIG-UHFFFAOYSA-N
Synonym 2-oxa-6-azaspiro 3.3 heptane oxalate, 2-oxa-6-aza-spiro 3.3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3,3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3.3 heptane xoxalate, 2-oxa-6-azaspiro 3.3 heptane oxalic acid, 2-oxa-6-azaspiro 3.3 heptane; oxalic acid, 2-oxa-6-azaspiro 3.3 heptane ethanedioate, 2-oxa-6-azaspiro 3.3 heptane oxalate 1:1, 2-oxa-6-azaspiro 3.3 heptane, ethanedioate 1:1, acmc-2099rj
PubChem CID 53308464
SMILES OC(=O)C(O)=O.C1NCC11COC1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
IUPAC Name 2-oxa-6-azaspiro[3.3]heptane; oxalic acid
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 2-Oxa-6-azaspiro[3.3]heptane oxalate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only