missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Octyne 99.0+%, TCI America™
$124.89 - $124.89
Chemical Identifiers
| CAS | 2809-67-8 |
|---|---|
| Molecular Formula | C8H14 |
| Molecular Weight (g/mol) | 110.20 |
| MDL Number | MFCD00027302 |
| InChI Key | QCQALVMFTWRCFI-UHFFFAOYSA-N |
| Synonym | 2-octyne, n-c5h11c.$.cch3, amylmethylacetylene, methylpentylacetylene, acmc-209h0u, n-c5h11c.equiv.cch3 |
| PubChem CID | 17769 |
| IUPAC Name | oct-2-yne |
| SMILES | CCCCCC#CC |
Chemical Identifiers
| 2809-67-8 | |
| 110.20 | |
| QCQALVMFTWRCFI-UHFFFAOYSA-N | |
| 17769 | |
| CCCCCC#CC |
| C8H14 | |
| MFCD00027302 | |
| 2-octyne, n-c5h11c.$.cch3, amylmethylacetylene, methylpentylacetylene, acmc-209h0u, n-c5h11c.equiv.cch3 | |
| oct-2-yne |
Specifications
| 2809-67-8 | |
| 137°C | |
| MFCD00027302 | |
| 3295 | |
| QCQALVMFTWRCFI-UHFFFAOYSA-N | |
| oct-2-yne | |
| 17769 | |
| ≥99.0% (GC) | |
| 2-Octyne |
| Colorless | |
| C8H14 | |
| 5 mL | |
| 2-octyne, n-c5h11c.$.cch3, amylmethylacetylene, methylpentylacetylene, acmc-209h0u, n-c5h11c.equiv.cch3 | |
| CCCCCC#CC | |
| 110.20 | |
| 110.20 | |
| Liquid |
Safety and Handling
TSCA : No