missing translation for 'onlineSavingsMsg'
Learn More

2-Octyn-1-ol, 98%

Catalog No. AAL0559609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0559609 10 g
AAL0559618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0559609 Supplier Thermo Scientific Chemicals Supplier No. L0559609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 20739-58-6 | C8H14O | 126.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20739-58-6
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00039542
InChI Key TTWYFVOMGMBZCF-UHFFFAOYSA-N
Synonym 2-octyn-1-ol, 2-octyne-1-ol, pubchem13066, acmc-1clfg, 2-octyn 1-ol, 2-octyn-1-ol 10g
PubChem CID 140750
IUPAC Name oct-2-yn-1-ol
SMILES CCCCCC#CCO

Specifications

Density 0.88
Boiling Point 76°C to 78°C (2 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.456
Quantity 10 g
Beilstein 1744120
Formula Weight 126.2
Percent Purity 98%
Chemical Name or Material 2-Octyn-1-ol
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only