missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Octenylsuccinic Anhydride (cis- and trans- mixture) 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Physical Form | Clear Liquid at 20°C |
| Formula Weight | 210.27 |
| Percent Purity | ≥95.0% (GC,T) |
| Chemical Name or Material | 2-Octenylsuccinic Anhydride (cis- and trans- mixture) |
Safety and Handling
| EINECSNumber | (2)-0852 |
| TSCA | Yes |