missing translation for 'onlineSavingsMsg'
Learn More

2-Octene (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. O004225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O004225ML 25 mL
O0042500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O004225ML Supplier TCI America Supplier No. O004225ML

Chemical Identifiers

CAS 111-67-1
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00009532
InChI Key ILPBINAXDRFYPL-HWKANZROSA-N
Synonym trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
PubChem CID 5364448
IUPAC Name (E)-oct-2-ene
SMILES CCCCCC=CC

Specifications

Color Colorless
Boiling Point 125°C
Quantity 25 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Octene (cis- and trans- mixture)
EINECSNumber (2)-0024
TSCA Yes