missing translation for 'onlineSavingsMsg'
Learn More

2-Octen-4-one, 97%

Catalog No. AAB2225006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2225006 5 g
AAB2225003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2225006 Supplier Thermo Scientific Chemicals Supplier No. B2225006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4643-27-0 | C8H14O | 126.199 g/mol

Reactant in the rhodium-catalyzed asymmetric ring opening of oxabicyclic alkenes with organoboronic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the rhodium-catalyzed asymmetric ring opening of oxabicyclic alkenes with organoboronic acids.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 4643-27-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00061023
InChI Key FMDLEUPBHMCPQV-GQCTYLIASA-N
Synonym 2-octen-4-one, propenyl butyl ketone, butyl propenyl ketone, unii-c9nb51lmxt, 2-octen-4-one natural, fema no. 3603, c9nb51lmxt, e-oct-2-en-4-one, 2e-oct-2-en-4-one, 2e-2-octen-4-one
PubChem CID 5365891
IUPAC Name (E)-oct-2-en-4-one
SMILES CCCCC(=O)C=CC

Specifications

Density 0.85
Boiling Point 65°C (15 mmHg)
Flash Point 58°C (136°F)
Refractive Index 1.445
Quantity 5 g
UN Number UN1224
Solubility Information Insoluble in water.
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 2-Octen-4-one
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 225-071-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only