Learn More
(+/-)-2-Octanol, 98%
Description
(±)-2-Octanol is used in flavors, paints and coatings, defoaming agent, low-volatility solvent, lubricants, fuels, paints and coatings. It is used as a frother in mineral floatation and wetting agent. It acts as an intermediate in the preparation of surfactants, cosmetic emollients esters and plasticizers. Further, it is also used as a reagent in the synthesis of pharmaceutical compounds and piperine derivatives. In addition to this, it is considered as an alternative of 2-ethylhexanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(^+)-2-Octanol is used in flavors, paints and coatings, defoaming agent, low-volatility solvent, lubricants, fuels, paints and coatings. It is used as a frother in mineral floatation and wetting agent. It acts as an intermediate in the preparation of surfactants, cosmetic emollients esters and plasticizers. Further, it is also used as a reagent in the synthesis of pharmaceutical compounds and piperine derivatives. In addition to this, it is considered as an alternative of 2-ethylhexanol.
Solubility
Miscible with diethyl ether, acetone, alcohol, most common organic solvents, aromatic and aliphatic hydrocarbons. Slightly miscible with cold water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -38°C |
| Density | 0.819 |
| Boiling Point | 179°C to 180°C |
| Flash Point | 76°C (169°F) |
| Odor | Pleasant |
| Refractive Index | 1.426 |
| Quantity | 100 mL |
| Beilstein | 1719325 |
| Merck Index | 14,6752 |
| Solubility Information | Miscible with diethyl ether,acetone,alcohol,most common organic solvents,aromatic and aliphatic hydrocarbons. Slightly miscible with cold water. |
| Show More |
Safety and Handling
| Hazard Category | H227-H318 |
| Hazard Statement | GHS H Statement H319-H227 Causes serious eye irritation. Combustible liquid. |
| Precautionary Statement | P210-P280-P305+P351+P338-P310-P370+P378q-P501c |
| EINECSNumber | 204-667-0 |
| RTECSNumber | RH0795000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only