missing translation for 'onlineSavingsMsg'
Learn More

2'-O-Methylcytidine 98.0+%, TCI America™

Catalog No. M2317200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2317200MG 200 mg
M23171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2317200MG Supplier TCI America Supplier No. M2317200MG

Chemical Identifiers

CAS 2140-72-9
Molecular Formula C10H15N3O5
Molecular Weight (g/mol) 257.246
MDL Number MFCD00056067
InChI Key RFCQJGFZUQFYRF-ZOQUXTDFSA-N
PubChem CID 150971
ChEBI CHEBI:19228
IUPAC Name 4-amino-1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]pyrimidin-2-one
SMILES COC1C(C(OC1N2C=CC(=NC2=O)N)CO)O

Specifications

Melting Point 257°C
Color White
Quantity 200 mg
Synonym 2'-o-methylcytidine, cytidine, 2'-o-methyl, 2'-o-methyl cytidine, o 2'-methylcytidine, 4-amino-1-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl pyrimidin-2 1h-one, 2'-o-methyl-cytidine, o-2-methylcytidine, o2'-methylcytidine, 2-o-methylcytidine, 2/'-o-methylcytidine
Formula Weight 257.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-O-Methylcytidine
TSCA No
Recommended Storage Refrigerator