missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-O-Methylcytidine, 99%

Catalog No. AAJ6552803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6552803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6552803 Supplier Thermo Scientific Chemicals Supplier No. J6552803

CAS: 2140-72-9 | C10H15N3O5 | 257.246 g/mol

2'-O-Methylcytidine is used in the preparation of nucleoside derivatives as inhibitors of RNA-dependent RNA viral polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-O-Methylcytidine is used in the preparation of nucleoside derivatives as inhibitors of RNA-dependent RNA viral polymerase.

Solubility
Soluble in DMSO.

Notes
Light & Air Sensitive

Chemical Identifiers

CAS 2140-72-9
Molecular Formula C10H15N3O5
Molecular Weight (g/mol) 257.246
MDL Number MFCD00056067
InChI Key RFCQJGFZUQFYRF-ZOQUXTDFSA-N
Synonym 2'-o-methylcytidine, cytidine, 2'-o-methyl, 2'-o-methyl cytidine, o 2'-methylcytidine, 4-amino-1-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl pyrimidin-2 1h-one, 2'-o-methyl-cytidine, o-2-methylcytidine, o2'-methylcytidine, 2-o-methylcytidine, 2/'-o-methylcytidine
PubChem CID 150971
ChEBI CHEBI:19228
IUPAC Name 4-amino-1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]pyrimidin-2-one
SMILES COC1C(C(OC1N2C=CC(=NC2=O)N)CO)O

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in DMSO.
Formula Weight 257.24
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-O-Methylcytidine
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only