missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-O-Methyladenosine, 99%

Catalog No. aaj6492203
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Catalog No. Quantity
AAJ6492203 1 g
1 options

Catalog No. AAJ6492203

Supplier: Thermo Scientific Chemicals J6492203

Only null left

CAS: 2140-79-6 | C11H15N5O4 | 281.27 g/mol

2'-O-Methyladenosine is an analog of adenosine used to prepare nucleoside derivatives as inhibitors of RNA-dependent RNA viral polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-O-Methyladenosine is an analog of adenosine used to prepare nucleoside derivatives as inhibitors of RNA-dependent RNA viral polymerase.

Solubility
Soluble in water.

Notes
Store away from st

Chemical Identifiers

CAS 2140-79-6
Molecular Formula C11H15N5O4
Molecular Weight (g/mol) 281.27
MDL Number MFCD00056002
InChI Key FPUGCISOLXNPPC-IOSLPCCCSA-N
Synonym 2'-o-methyladenosine, adenosine, 2'-o-methyl, 2'-o-methyl adenosine, 2r,3r,4r,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl-4-methoxytetrahydrofuran-3-ol, unii-02yx82ihz5, chembl73237, 2'-o-methyl-adenosine, 2r,3r,4r,5r-5-6-aminopurin-9-yl-2-hydroxymethyl-4-methoxyoxolan-3-ol, 2r,3r,4r,5r-5-6-aminopurin-9-yl-2-hydroxymethyl-4-methoxy-tetrahydrofuran-3-ol, cordysinin b
PubChem CID 102213
ChEBI CHEBI:69426
IUPAC Name (2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)-4-methoxyoxolan-3-ol
SMILES CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C(N)N=CN=C12

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 281.27
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-O-Methyladenosine
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only