missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Norbornene 99.0+%, TCI America™
$39.14 - $143.54
Chemical Identifiers
| CAS | 498-66-8 |
|---|---|
| Molecular Formula | C7H10 |
| Molecular Weight (g/mol) | 94.157 |
| MDL Number | MFCD00082304 |
| InChI Key | JFNLZVQOOSMTJK-UHFFFAOYSA-N |
| Synonym | bicyclo 2.2.1 hept-2-ene, norbornylene, 2-norbornene, norbornene, norcamphene, norfenchene, 2-norbornylene, 3,6-endomethylenecyclohexene, bicyclo 2.2.1-2-heptene, bicyclo 2.2.1.-2-heptene |
| PubChem CID | 10352 |
| IUPAC Name | bicyclo[2.2.1]hept-2-ene |
| SMILES | C1CC2CC1C=C2 |
Chemical Identifiers
| 498-66-8 | |
| 94.157 | |
| JFNLZVQOOSMTJK-UHFFFAOYSA-N | |
| 10352 | |
| C1CC2CC1C=C2 |
| C7H10 | |
| MFCD00082304 | |
| bicyclo 2.2.1 hept-2-ene, norbornylene, 2-norbornene, norbornene, norcamphene, norfenchene, 2-norbornylene, 3,6-endomethylenecyclohexene, bicyclo 2.2.1-2-heptene, bicyclo 2.2.1.-2-heptene | |
| bicyclo[2.2.1]hept-2-ene |
Specifications
| 498-66-8 | |
| White | |
| C7H10 | |
| 25 g | |
| bicyclo 2.2.1 hept-2-ene, norbornylene, 2-norbornene, norbornene, norcamphene, norfenchene, 2-norbornylene, 3,6-endomethylenecyclohexene, bicyclo 2.2.1-2-heptene, bicyclo 2.2.1.-2-heptene | |
| C1CC2CC1C=C2 | |
| 94.157 | |
| 94.16 | |
| Crystalline Powder |
| 47°C | |
| 96°C | |
| MFCD00082304 | |
| 1325 | |
| JFNLZVQOOSMTJK-UHFFFAOYSA-N | |
| bicyclo[2.2.1]hept-2-ene | |
| 10352 | |
| ≥99.0% (GC) | |
| 2-Norbornene |
Safety and Handling
EINECSNumber : (4)-1763
RTECSNumber : RB7900000
TSCA : Yes