missing translation for 'onlineSavingsMsg'
Learn More

2-Norbornene 99.0+%, TCI America™

Catalog No. N016625G
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N016625G 25 g
N0166400G 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N016625G Supplier TCI America Supplier No. N016625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 498-66-8
Molecular Formula C7H10
Molecular Weight (g/mol) 94.157
MDL Number MFCD00082304
InChI Key JFNLZVQOOSMTJK-UHFFFAOYSA-N
Synonym bicyclo 2.2.1 hept-2-ene, norbornylene, 2-norbornene, norbornene, norcamphene, norfenchene, 2-norbornylene, 3,6-endomethylenecyclohexene, bicyclo 2.2.1-2-heptene, bicyclo 2.2.1.-2-heptene
PubChem CID 10352
IUPAC Name bicyclo[2.2.1]hept-2-ene
SMILES C1CC2CC1C=C2

Specifications

Melting Point 47°C
Color White
Boiling Point 96°C
Quantity 25 g
UN Number 1325
Formula Weight 94.16
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Norbornene
EINECSNumber (4)-1763
RTECSNumber RB7900000
TSCA Yes