missing translation for 'onlineSavingsMsg'
Learn More

2-Nonyn-1-ol, 96%

Catalog No. AAL0638006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0638006 5 g
AAL0638014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0638006 Supplier Thermo Scientific Chemicals Supplier No. L0638006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5921-73-3 | C9H16O | 140.23 g/mol

2-Nonyn-1-ol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nonyn-1-ol is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5921-73-3
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00041566
InChI Key XERDOEKKHDALKJ-UHFFFAOYSA-N
Synonym 2-nonyn-1-ol, pubchem13067, acmc-1apv0
PubChem CID 80017
IUPAC Name non-2-yn-1-ol
SMILES CCCCCCC#CCO

Specifications

Density 0.873
Boiling Point 109°C to 111°C (11 mmHg)
Refractive Index 1.456
Quantity 5 g
Beilstein 1745753
Solubility Information Not miscible in water.
Formula Weight 140.23
Percent Purity 96%
Chemical Name or Material 2-Nonyn-1-ol
EINECSNumber 227-646-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only