missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nonanone 98.0+%, TCI America™
$39.14 - $218.10
Chemical Identifiers
| CAS | 821-55-6 |
|---|---|
| Molecular Formula | C9H18O |
| Molecular Weight (g/mol) | 142.242 |
| MDL Number | MFCD00009553 |
| InChI Key | VKCYHJWLYTUGCC-UHFFFAOYSA-N |
| Synonym | 2-nonanone, heptyl methyl ketone, methyl heptyl ketone, beta-nonanone, ketone, heptyl methyl, methyl n-heptyl ketone, 2-nonanone natural, nonanone, .beta.-nonanone, unii-ze5k73yn2z |
| PubChem CID | 13187 |
| ChEBI | CHEBI:77927 |
| IUPAC Name | nonan-2-one |
| SMILES | CCCCCCCC(=O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
N029325ML
|
TCI America
N029325ML |
25 mL |
Each for $39.14
|
|
|||||
|
N0293500ML
|
TCI America
N0293500ML |
500 mL |
Each for $218.10
|
|
|||||
Chemical Identifiers
| 821-55-6 | |
| 142.242 | |
| VKCYHJWLYTUGCC-UHFFFAOYSA-N | |
| 13187 | |
| nonan-2-one |
| C9H18O | |
| MFCD00009553 | |
| 2-nonanone, heptyl methyl ketone, methyl heptyl ketone, beta-nonanone, ketone, heptyl methyl, methyl n-heptyl ketone, 2-nonanone natural, nonanone, .beta.-nonanone, unii-ze5k73yn2z | |
| CHEBI:77927 | |
| CCCCCCCC(=O)C |
Specifications
| 821-55-6 | |
| Yellow | |
| C9H18O | |
| 25 mL | |
| VKCYHJWLYTUGCC-UHFFFAOYSA-N | |
| nonan-2-one | |
| 13187 | |
| 142.24 | |
| Liquid |
| -21°C | |
| 195°C | |
| MFCD00009553 | |
| 2-nonanone, heptyl methyl ketone, methyl heptyl ketone, beta-nonanone, ketone, heptyl methyl, methyl n-heptyl ketone, 2-nonanone natural, nonanone, .beta.-nonanone, unii-ze5k73yn2z | |
| CCCCCCCC(=O)C | |
| 142.242 | |
| CHEBI:77927 | |
| ≥98.0% (GC) | |
| 2-Nonanone |
Safety and Handling
EINECSNumber : (2)-0542
RTECSNumber : RA8225000
TSCA : Yes