missing translation for 'onlineSavingsMsg'
Learn More

2-Nonanone, 97%, Thermo Scientific Chemicals

CAS: 821-55-6 | C9H18O | 142.24 g/mol

$105.50 - $403.83

Chemical Identifiers

CAS 821-55-6
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00009553
InChI Key VKCYHJWLYTUGCC-UHFFFAOYSA-N
Synonym 2-nonanone, heptyl methyl ketone, methyl heptyl ketone, beta-nonanone, ketone, heptyl methyl, methyl n-heptyl ketone, 2-nonanone natural, nonanone, .beta.-nonanone, unii-ze5k73yn2z
PubChem CID 13187
ChEBI CHEBI:77927
IUPAC Name nonan-2-one
SMILES CCCCCCCC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC148781000
SDS View Documents
Thermo Scientific Chemicals
148781000
100 mL Glass bottle
Each for $105.50
Only null left
 
AC148785000
SDS View Documents
Thermo Scientific Chemicals
148785000
500 mL Glass Bottle
Each for $403.83
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

821-55-6
142.24
VKCYHJWLYTUGCC-UHFFFAOYSA-N
13187
nonan-2-one
C9H18O
MFCD00009553
2-nonanone, heptyl methyl ketone, methyl heptyl ketone, beta-nonanone, ketone, heptyl methyl, methyl n-heptyl ketone, 2-nonanone natural, nonanone, .beta.-nonanone, unii-ze5k73yn2z
CHEBI:77927
CCCCCCCC(=O)C

Specifications

821-55-6
100.0
Yellow
188.0°C to 190.0°C
Authentic
Glass bottle
1.4200 to 1.4220
MFCD00009553
01, 709
2-nonanone, heptyl methyl ketone, methyl heptyl ketone, beta-nonanone, ketone, heptyl methyl, methyl n-heptyl ketone, 2-nonanone natural, nonanone, .beta.-nonanone, unii-ze5k73yn2z
VKCYHJWLYTUGCC-UHFFFAOYSA-N
nonan-2-one
13187
142.24
Liquid
98.5
-21.0°C
0.8260g/mL
68°C
98.5% min. (GC)
C9H18O
CH3(CH2)6COCH3
100 mL
0.826
Solubility in water: 0.37g/L (25°C). Other solubilities: soluble in acetone
CCCCCCCC(=O)C
142.24
CHEBI:77927
99%
2-Nonanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 212-480-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only