missing translation for 'onlineSavingsMsg'
Learn More

2-Nonanol 98.0+%, TCI America™

Catalog No. p-7133756
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N033425ML 25 mL
N0334500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N033425ML Supplier TCI America Supplier No. N033425ML
Only null left

Chemical Identifiers

CAS 628-99-9
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.258
MDL Number MFCD00004593
InChI Key NGDNVOAEIVQRFH-UHFFFAOYSA-N
Synonym 2-nonanol, 1-methyl-1-octanol, 2-nonyl alcohol, methyl heptyl carbinol, 2-hydroxynonane, heptylmethylcarbinol, n-nonan-2-ol, nonanol-2, heptyl methyl carbinol, 2-nonanol natural
PubChem CID 12367
ChEBI CHEBI:78304
IUPAC Name nonan-2-ol
SMILES CCCCCCCC(C)O

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 mL
Formula Weight 144.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Nonanol
EINECSNumber (2)-0217
TSCA Yes