missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$100.11 - $100.11
Chemical Identifiers
| CAS | 5570-19-4 |
|---|---|
| Molecular Formula | C6H6BNO4 |
| Molecular Weight (g/mol) | 166.93 |
| MDL Number | MFCD00161358 |
| InChI Key | SFUIGUOONHIVLG-UHFFFAOYSA-N |
| PubChem CID | 2773548 |
| IUPAC Name | (2-nitrophenyl)boronic acid |
| SMILES | OB(O)C1=CC=CC=C1[N+]([O-])=O |
Chemical Identifiers
| 5570-19-4 | |
| 166.93 | |
| SFUIGUOONHIVLG-UHFFFAOYSA-N | |
| (2-nitrophenyl)boronic acid |
| C6H6BNO4 | |
| MFCD00161358 | |
| 2773548 | |
| OB(O)C1=CC=CC=C1[N+]([O-])=O |
Specifications
| 5570-19-4 | |
| C6H6BNO4 | |
| 1 g | |
| SFUIGUOONHIVLG-UHFFFAOYSA-N | |
| (2-nitrophenyl)boronic acid | |
| 2773548 | |
| Crystalline Powder |
| 142°C | |
| MFCD00161358 | |
| 2-nitrophenyl boronic acid, 2-nitrobenzeneboronic acid, boronic acid, nitrophenyl, o-nitrophenylboronic acid, 2-borononitrobenzene, pubchem8087, nitrobenzeneboronic acid, pubchem16455, 2-nitrophenylboronicacid | |
| OB(O)C1=CC=CC=C1[N+]([O-])=O | |
| 166.93 | |
| 166.93 | |
| 2-Nitrophenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No