missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenylacetic acid, 99%, Thermo Scientific™

Catalog No. AC157060250
Change view
Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157060250 25g Glass bottle
AC157061000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC157060250 Supplier Thermo Scientific Chemicals Supplier No. 157060250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 3740-52-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007190
InChI Key WMUZDBZPDLHUMW-UHFFFAOYSA-N
Synonym 2-nitrophenylacetic acid, 2-2-nitrophenyl acetic acid, 2-nitrophenyl acetic acid, o-nitrophenylacetic acid, benzeneacetic acid, 2-nitro, acetic acid, o-nitrophenyl, ortho-nitrophenyl acetic acid, 2-o-nitrophenyl acetic acid, 2-nitrobenzeneacetic acid, unii-05tn0suy38
PubChem CID 77337
IUPAC Name 2-(2-nitrophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC=C1[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula O2NC6H4CH2CO2H
Quantity 25g
Beilstein 09, 454
Formula Weight 181.15
Percent Purity 99%
Chemical Name or Material 2-Nitrophenylacetic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-128-0
RTECSNumber AJ1130000
TSCA TSCA

RUO – Research Use Only