missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenyl isocyanate, 99%

Catalog No. AC296690050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296690050 5 g Glass bottle
AC296690250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC296690050 Supplier Thermo Scientific Chemicals Supplier No. 296690050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3320-86-3 | C7H4N2O3 | 164.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3320-86-3
Molecular Formula C7H4N2O3
Molecular Weight (g/mol) 164.12
InChI Key JRVZITODZAQRQM-UHFFFAOYSA-N
Synonym 2-nitrophenyl isocyanate, o-nitrophenyl isocyanate, benzene, 1-isocyanato-2-nitro, isocyanic acid, o-nitrophenyl ester, 2-nitrophenylisocyanate, 2-nitrobenzenisocyanate, nitrophenyl isocyanate, 2-nitrophenylisocynate, 2-nitro phenyl isocyanate, acmc-1cp5k
PubChem CID 76834
IUPAC Name 1-isocyanato-2-nitrobenzene
SMILES C1=CC=C(C(=C1)N=C=O)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 38.0°C to 41.0°C
Color Yellow
Boiling Point 135.0°C to 137.0°C (17.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4NCO
Quantity 5 g
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in toluene
Formula Weight 164.12
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 2-Nitrophenyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
EINECSNumber 222-024-2

RUO – Research Use Only