missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenyl-beta-D-galactopyranoside, 98+%

Catalog No. p-7045494
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1093903 1 g
AAL1093906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1093903 Supplier Thermo Scientific Chemicals Supplier No. L1093903
Only null left

CAS: 369-07-3 | C12H15NO8 | 301.251 g/mol

2-Nitrophenyl-beta-D-galactopyranoside is a β-Galactosidase substrate for colorimetric and EIA applications; counterpart of widely employed pNPP/alkaline phosphatase substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrophenyl-beta-D-galactopyranoside is a β-Galactosidase substrate for colorimetric and EIA applications; counterpart of widely employed pNPP/alkaline phosphatase substrate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 20°C. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 369-07-3
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.251
MDL Number MFCD00063255
InChI Key KUWPCJHYPSUOFW-YBXAARCKSA-N
Synonym 2-nitrophenyl-beta-d-galactopyranoside, onpg, 2-nitrophenyl beta-d-galactopyranoside, 2-nitrophenyl beta-d-galactoside, o-nitrophenyl beta-d-galactopyranoside, o-nitrophenyl-beta-d-galactoside, 1-o-o-nitrophenyl-beta-d-galactopyranose, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2-nitrophenoxy oxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2-nitrophenoxy tetrahydro-2h-pyran-3,4,5-triol, 2-nitrophenyl b-d-galactopyranoside
PubChem CID 96647
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(2-nitrophenoxy)oxane-3,4,5-triol
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point ∽192°C (decomposition)
Quantity 1 g
Beilstein 92207
Solubility Information Soluble in water.
Optical Rotation −68° (c=1 in Water)
Formula Weight 301.25
Percent Purity ≥98%
Chemical Name or Material 2-Nitrophenyl-beta-D-galactopyranoside
EINECSNumber 206-716-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only