missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroimidazole 98.0+%, TCI America™

Catalog No. N08915G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08915G 5 g
N08911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08915G Supplier TCI America Supplier No. N08915G

Chemical Identifiers

CAS 527-73-1
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00005185
InChI Key YZEUHQHUFTYLPH-UHFFFAOYSA-N
Synonym 2-nitroimidazole, azomycin, amicin, nitroimidazole, 1h-imidazole, 2-nitro, imidazole, 2-nitro, 2-nitroimidazol, 2-nitro-imidazole, unii-k8e96xl55d, azomycin 2-nitroimidazole
PubChem CID 10701
ChEBI CHEBI:67135
IUPAC Name 2-nitro-1H-imidazole
SMILES [O-][N+](=O)C1=NC=CN1

Specifications

Melting Point 288°C
Color Yellow
Quantity 5 g
UN Number 2811
Formula Weight 113.08
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitroimidazole
RTECSNumber NI7875000
TSCA Yes