missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroimidazole, 98%

Catalog No. p-7046233
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13546MC 100 mg
AAL13546ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL13546MC Supplier Thermo Scientific Chemicals Supplier No. L13546MC

CAS: 527-73-1 | C3H3N3O2 | 113.08 g/mol

2-Nitroimidazole, is used as an antibiotic in pharma industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitroimidazole, is used as an antibiotic in pharma industry.

Solubility
It is slightly soluble in water. Soluble in acetone, butyl acetate, ethanol, ethyl acetate and methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids, acid chlorides, anhydrous acids.. Stable under recommended storage conditions.

Chemical Identifiers

CAS 527-73-1
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00005185
InChI Key YZEUHQHUFTYLPH-UHFFFAOYSA-N
Synonym 2-nitroimidazole, azomycin, amicin, nitroimidazole, 1h-imidazole, 2-nitro, imidazole, 2-nitro, 2-nitroimidazol, 2-nitro-imidazole, unii-k8e96xl55d, azomycin 2-nitroimidazole
PubChem CID 10701
ChEBI CHEBI:67135
IUPAC Name 2-nitro-1H-imidazole
SMILES [O-][N+](=O)C1=NC=CN1

Specifications

Melting Point ∽287°C (decomposition)
Quantity 100 mg
UN Number UN2811
Beilstein 116444
Merck Index 14,921
Solubility Information It is slightly soluble in water. Soluble in acetone,butyl acetate,ethanol,ethyl acetate and methanol.
Formula Weight 113.08
Percent Purity 98%
Chemical Name or Material 2-Nitroimidazole
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 208-425-5
RTECSNumber NI7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only