missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroimidazole, 98%

CAS: 527-73-1 | C3H3N3O2 | 113.08 g/mol

$565.58 - $565.58

Chemical Identifiers

CAS 527-73-1
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00005185
InChI Key YZEUHQHUFTYLPH-UHFFFAOYSA-N
Synonym 2-nitroimidazole, azomycin, amicin, nitroimidazole, 1h-imidazole, 2-nitro, imidazole, 2-nitro, 2-nitroimidazol, 2-nitro-imidazole, unii-k8e96xl55d, azomycin 2-nitroimidazole
PubChem CID 10701
ChEBI CHEBI:67135
IUPAC Name 2-nitro-1H-imidazole
SMILES [O-][N+](=O)C1=NC=CN1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC187570010
SDS View Documents
Thermo Scientific Chemicals
187570010
1 g Glass bottle
Each for $565.58
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

527-73-1
113.08
YZEUHQHUFTYLPH-UHFFFAOYSA-N
10701
2-nitro-1H-imidazole
C3H3N3O2
MFCD00005185
2-nitroimidazole, azomycin, amicin, nitroimidazole, 1h-imidazole, 2-nitro, imidazole, 2-nitro, 2-nitroimidazol, 2-nitro-imidazole, unii-k8e96xl55d, azomycin 2-nitroimidazole
CHEBI:67135
[O-][N+](=O)C1=NC=CN1

Specifications

527-73-1
100.0
Green-Khaki to Yellow
98%
C3H3N3O2
1 g
2-nitroimidazole, azomycin, amicin, nitroimidazole, 1h-imidazole, 2-nitro, imidazole, 2-nitro, 2-nitroimidazol, 2-nitro-imidazole, unii-k8e96xl55d, azomycin 2-nitroimidazole
YZEUHQHUFTYLPH-UHFFFAOYSA-N
2-nitro-1H-imidazole
10701
113.08
Powder
97.5
287°C
Authentic
Glass bottle
MFCD00005185
15, 914
Solubility in water: insoluble in water. Other solubilities: soluble in alkaline h2o- insoluble in acidic h2o,soluble in methanol,ethanol,acetone,ethyl,acetate and butyl acetate,practically insoluble in ether and chloroform
[O-][N+](=O)C1=NC=CN1
113.08
CHEBI:67135
98%
2-Nitroimidazole, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye prot

GHS Signal Word: Danger

EINECSNumber : 208-425-5

RTECSNumber : NI7875000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only