missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroethanol, tech. 80%

Catalog No. aaa1768122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1768122 100 g
AAA1768106 5 g
AAA1768114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1768122 Supplier Thermo Scientific Chemicals Supplier No. A1768122
Only null left

CAS: 625-48-9 | C2H5NO3 | 91.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 625-48-9
Molecular Formula C2H5NO3
Molecular Weight (g/mol) 91.066
MDL Number MFCD00007405
InChI Key KIPMDPDAFINLIV-UHFFFAOYSA-N
Synonym ethanol, 2-nitro, 2-nitroethan-1-ol, beta-nitroalcohol, beta-nitroethanol, 2-nitro-ethanol, .beta.-nitroethanol, ccris 6076, 2 nitroethanol, pubchem14208, 2-nitroethanol
PubChem CID 12252
IUPAC Name 2-nitroethanol
SMILES C(CO)[N+](=O)[O-]

Specifications

Melting Point -80°C
Density 1.27
Boiling Point 194°C
Flash Point >110°C (230°F)
Refractive Index 1.4451
Quantity 100 g
Beilstein 1633753
Formula Weight 91.07
Percent Purity 80%
Grade Technical
Chemical Name or Material 2-Nitroethanol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-895-1
RTECSNumber KL9370000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only