missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 2-Nitroethanol

Catalog No. 146633100G
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
146633100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 146633100G Supplier Sigma Aldrich Supplier No. 146633100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Nitroethanol

Specifications

CAS 625-48-9
Melting Point -80°C (lit.)
Density 1.27 g/mL (at 25°C (literature))
Boiling Point 194°C (765 mmHg)
Molecular Formula C2H5NO3
Refractive Index n20/D 1.445 (literature)
Linear Formula NO2CH2CH2OH
MDL Number MFCD00007405
Quantity 100 g
Molecular Weight (g/mol) 91.07
Percent Purity 97%
Show More Show Less