missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzenesulfonyl chloride, 97%

CAS: 1694-92-4 | C6H4ClNO4S | 221.61 g/mol

$175.44 - $175.44

Chemical Identifiers

CAS 1694-92-4
Molecular Formula C6H4ClNO4S
Molecular Weight (g/mol) 221.61
MDL Number MFCD00007430
InChI Key WPHUUIODWRNJLO-UHFFFAOYSA-N
Synonym 2-nitrobenzene-1-sulfonyl chloride, o-nitrobenzenesulfonyl chloride, o-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, 2-nitro, 2-nitrobenzenesulphonyl chloride, 2-nitrobenzenesulfonylchloride, 2-nitro-benzenesulfonyl chloride, benzenesulfonyl chloride, o-nitro, 2-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, nitro
PubChem CID 15540
IUPAC Name 2-nitrobenzenesulfonyl chloride
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC128431000
SDS View Documents
Thermo Scientific Chemicals
128431000
100 g Glass bottle
Each for $175.44
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1694-92-4
221.61
WPHUUIODWRNJLO-UHFFFAOYSA-N
15540
C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)Cl
C6H4ClNO4S
MFCD00007430
2-nitrobenzene-1-sulfonyl chloride, o-nitrobenzenesulfonyl chloride, o-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, 2-nitro, 2-nitrobenzenesulphonyl chloride, 2-nitrobenzenesulfonylchloride, 2-nitro-benzenesulfonyl chloride, benzenesulfonyl chloride, o-nitro, 2-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, nitro
2-nitrobenzenesulfonyl chloride

Specifications

1694-92-4
96% min. (GC)
C6H4ClNO4S
MFCD00007430
11, 67
2-nitrobenzene-1-sulfonyl chloride, o-nitrobenzenesulfonyl chloride, o-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, 2-nitro, 2-nitrobenzenesulphonyl chloride, 2-nitrobenzenesulfonylchloride, 2-nitro-benzenesulfonyl chloride, benzenesulfonyl chloride, o-nitro, 2-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, nitro
C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)Cl
221.61
221.61
2-Nitrobenzenesulfonyl chloride
63.0°C to 67.0°C
Glass bottle
SO2ClC6H4NO2
100 g
06,419
WPHUUIODWRNJLO-UHFFFAOYSA-N
2-nitrobenzenesulfonyl chloride
15540
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 216-907-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only