missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzenesulfonyl chloride, 97%

Catalog No. AAB2152214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2152214 25 g
AAB2152222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2152214 Supplier Thermo Scientific Chemicals Supplier No. B2152214

CAS: 1694-92-4 | C6H4ClNO4S | 221.611 g/mol

2-Nitrobenzenesulfonyl chloride acts as a reagent to form 2-nitrobenzenesulfonyl derivatives of alcohols and amines. It is also involved in the selective cleavage with thiophenol without displacement of the nitro group. Further, it is used in the preparation of renin inhibitors. In addition to this, it finds application as an anticonvulsant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrobenzenesulfonyl chloride acts as a reagent to form 2-nitrobenzenesulfonyl derivatives of alcohols and amines. It is also involved in the selective cleavage with thiophenol without displacement of the nitro group. Further, it is used in the preparation of renin inhibitors. In addition to this, it finds application as an anticonvulsant.

Solubility
Soluble in toluene, tetrahydrofuran, methylene chloride, ethyl acetate and dimethylformamide. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 1694-92-4
Molecular Formula C6H4ClNO4S
Molecular Weight (g/mol) 221.611
MDL Number MFCD00007430
InChI Key WPHUUIODWRNJLO-UHFFFAOYSA-N
Synonym 2-nitrobenzene-1-sulfonyl chloride, o-nitrobenzenesulfonyl chloride, o-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, 2-nitro, 2-nitrobenzenesulphonyl chloride, 2-nitrobenzenesulfonylchloride, 2-nitro-benzenesulfonyl chloride, benzenesulfonyl chloride, o-nitro, 2-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, nitro
PubChem CID 15540
IUPAC Name 2-nitrobenzenesulfonyl chloride
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)Cl

Specifications

Melting Point 64°C to 69°C
Assay Percent Range 0.97
Quantity 25 g
UN Number UN3261
Beilstein 521155
Sensitivity Moisture sensitive
Solubility Information Soluble in toluene,tetrahydrofuran,methylene chloride,ethyl acetate and dimethylformamide. Insoluble in water.
Formula Weight 221.62
Percent Purity 97%
Chemical Name or Material 2-Nitrobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 216-907-1
TSCA Yes
storageNote1 Moisture Sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only