missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzeneboronic acid, 96%

Catalog No. AAL1798803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1798803 1 g
AAL1798806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1798803 Supplier Thermo Scientific Chemicals Supplier No. L1798803
Only null left

CAS: 5570-19-4 | C6H6BNO4 | 166.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5570-19-4
Molecular Formula C6H6BNO4
Molecular Weight (g/mol) 166.93
MDL Number MFCD00161358
InChI Key SFUIGUOONHIVLG-UHFFFAOYSA-N
Synonym 2-nitrophenyl boronic acid, 2-nitrobenzeneboronic acid, boronic acid, nitrophenyl, o-nitrophenylboronic acid, 2-borononitrobenzene, pubchem8087, nitrobenzeneboronic acid, pubchem16455, 2-nitrophenylboronicacid
PubChem CID 2773548
IUPAC Name (2-nitrophenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 149°C to 151°C
Quantity 1 g
Beilstein 3032097
Formula Weight 166.93
Percent Purity 96%
Chemical Name or Material 2-Nitrobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only