missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzamide 98.0+%, TCI America™

Catalog No. N032225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N032225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N032225G Supplier TCI America Supplier No. N032225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 610-15-1
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.136
MDL Number MFCD00007976
InChI Key KLGQWSOYKYFBTR-UHFFFAOYSA-N
Synonym o-nitrobenzamide, benzamide, o-nitro, benzamide, 2-nitro, 2-carbamoylnitrobenzene, nitrobenzamide, nitro-benzamide, 2-nitro-benzamide, benzamide,2-nitro, benzmide, 2-nitro, wln: zvr bnw
PubChem CID 11876
IUPAC Name 2-nitrobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)[N+](=O)[O-]

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 25 g
Formula Weight 166.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzamide
RTECSNumber CV5600000
TSCA No