missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzamide, 98%

Catalog No. AAA1408418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1408418 50 g
AAA1408409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1408418 Supplier Thermo Scientific Chemicals Supplier No. A1408418
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 610-15-1 | C7H6N2O3 | 166.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 610-15-1
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.136
MDL Number MFCD00007976
InChI Key KLGQWSOYKYFBTR-UHFFFAOYSA-N
Synonym o-nitrobenzamide, benzamide, o-nitro, benzamide, 2-nitro, 2-carbamoylnitrobenzene, nitrobenzamide, nitro-benzamide, 2-nitro-benzamide, benzamide,2-nitro, benzmide, 2-nitro, wln: zvr bnw
PubChem CID 11876
IUPAC Name 2-nitrobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)[N+](=O)[O-]

Specifications

Melting Point 174°C to 178°C
Boiling Point 317°C
Quantity 50 g
Beilstein 1950928
Formula Weight 166.14
Percent Purity 98%
Chemical Name or Material 2-Nitrobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-208-5
RTECSNumber CV5600000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only