missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroaniline 99.0+%, TCI America™

Catalog No. N011825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N011825G 25 g
N0118500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N011825G Supplier TCI America Supplier No. N011825G

Chemical Identifiers

CAS 88-74-4
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00007687
InChI Key DPJCXCZTLWNFOH-UHFFFAOYSA-N
Synonym o-nitroaniline, benzenamine, 2-nitro, o-nitraniline, 1-amino-2-nitrobenzene, 2-nitrobenzenamine, aniline, o-nitro, devol orange b, orange grs salt, o-aminonitrobenzene, 2-aminonitrobenzene
PubChem CID 6946
IUPAC Name 2-nitroaniline
SMILES C1=CC=C(C(=C1)N)[N+](=O)[O-]

Specifications

Melting Point 73°C
Boiling Point 165°C
Quantity 25 g
UN Number 1661
Formula Weight 138.13
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitroaniline
EINECSNumber (3)-0392&(5)-2301
RTECSNumber BY6650000
TSCA Yes