missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroamino-2-imidazoline, TCI America™

Catalog No. N08585G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08585G Supplier TCI America Supplier No. N08585G
Only null left

Chemical Identifiers

CAS 5465-96-3
Molecular Formula C3H6N4O2
Molecular Weight (g/mol) 130.107
MDL Number MFCD00047040
InChI Key DJZWNSRUEJSEEB-UHFFFAOYSA-N
Synonym 2-(Nitroimino)imidazolidine
PubChem CID 410042
IUPAC Name N-(4,5-dihydro-1H-imidazol-2-yl)nitramide
SMILES C1CN=C(N1)N[N+](=O)[O-]

Specifications

Melting Point 222°C
Color White
Quantity 5 g
Formula Weight 130.11
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitroamino-2-imidazoline