missing translation for 'onlineSavingsMsg'
Learn More

2'-Nitroacetophenone 96.0+%, TCI America™

Catalog No. p-7133624
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N01105G 5 g
N011025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N01105G Supplier TCI America Supplier No. N01105G
Only null left

Chemical Identifiers

CAS 577-59-3
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007144
InChI Key SUGXZLKUDLDTKX-UHFFFAOYSA-N
Synonym 2'-nitroacetophenone, 1-2-nitrophenyl ethanone, o-nitroacetophenone, ethanone, 1-2-nitrophenyl, 2-acetylnitrobenzene, acetophenone, 2'-nitro, methyl 2-nitrophenyl ketone, unii-52l53k4x3o, ccris 2329, 1-2-nitrophenyl ethan-1-one
PubChem CID 11346
IUPAC Name 1-(2-nitrophenyl)ethan-1-one
SMILES CC(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Color Yellow
Boiling Point 179°C
Quantity 5 g
Formula Weight 165.15
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2′-Nitroacetophenone
RTECSNumber AM9600000
TSCA Yes