missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-1-propanol, 97%

Catalog No. p-7059040
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4384306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4384306 Supplier Thermo Scientific Chemicals Supplier No. 04384306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2902-96-7 | C3H7NO3 | 105.09 g/mol

2-Nitro-1-propanol is used in improved synthesis of 2,2,2-trinitroethyl ethers, inhibits methane production during in vitro ruminal fermentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2902-96-7
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.09
MDL Number MFCD00007398
InChI Key PCNWBUOSTLGPMI-UHFFFAOYNA-N
Synonym 2-nitro-1-propanol, 2-nitropropanol, 1-propanol, 2-nitro, 2-nitropropanol-1, ccris 5045, 2-nitro-propan-1-ol, acmc-20anp9, 1-hydroxy-2-nitropropane, 1-propanol, 2-nitro-8ci
PubChem CID 92187
IUPAC Name 2-nitropropan-1-ol
SMILES CC(CO)[N+]([O-])=O

Specifications

Density 1.185 g/mL
Boiling Point 73°C (1 mmHg)
Flash Point 100°C (212°F)
Refractive Index 1.439
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 105.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Nitro-1-propanol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only