missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthylacetonitrile 98.0+%, TCI America™

Catalog No. N08745G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08745G 5 g
N087425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08745G Supplier TCI America Supplier No. N08745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7498-57-9
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.21
MDL Number MFCD00004122
InChI Key LPCWDVLDJVZIHA-UHFFFAOYSA-N
Synonym 2-naphthylacetonitrile, 2-naphthalen-2-yl acetonitrile, 2-naphthaleneacetonitrile, 2-naphthyl acetonitrile, naphthalene-2-acetonitrile, 2-2-naphthyl acetonitrile, .beta.-naphthyleneacetonitrile, 2-naphtylacetonitrile, 2-napthylacetonitrile, 2-naphthylmethylcyanide
PubChem CID 82008
IUPAC Name 2-(naphthalen-2-yl)acetonitrile
SMILES N#CCC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 86°C
Color White-Yellow
Boiling Point 303°C
Quantity 5 g
UN Number 3439
Formula Weight 167.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthylacetonitrile