Learn More
2-Naphthylacetonitrile, 97%
CAS: 7498-57-9 | C12H9N | 167.21 g/mol
Supplier: Thermo Scientific Chemicals L0938414
Description
2-Naphthylacetonitrile was used as starting reagent in the synthesis of 2-(1-cyano-1-(2-naphthyl))methylidene-3-phenyl-thiazolidine-4,5-dione.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Naphthylacetonitrile was used as starting reagent in the synthesis of 2-(1-cyano-1-(2-naphthyl))methylidene-3-phenyl-thiazolidine-4,5-dione.
Solubility
Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 2-Naphthylacetonitrile | |
| 82°C to 84°C | |
| 303°C | |
| MFCD00004122 | |
| UN3439 | |
| 2-naphthylacetonitrile, 2-naphthalen-2-yl acetonitrile, 2-naphthaleneacetonitrile, 2-naphthyl acetonitrile, naphthalene-2-acetonitrile, 2-2-naphthyl acetonitrile, .beta.-naphthyleneacetonitrile, 2-naphtylacetonitrile, 2-napthylacetonitrile, 2-naphthylmethylcyanide | |
| LPCWDVLDJVZIHA-UHFFFAOYSA-N | |
| 2-naphthalen-2-ylacetonitrile | |
| 82008 | |
| 97% |
| 7498-57-9 | |
| 1.092 | |
| C12H9N | |
| 25 g | |
| 1862879 | |
| Insoluble in water. | |
| N#CCC1=CC=C2C=CC=CC2=C1 | |
| 167.21 | |
| 167.21 |
Chemical Identifiers
| 7498-57-9 | |
| 167.21 | |
| LPCWDVLDJVZIHA-UHFFFAOYSA-N | |
| 82008 | |
| N#CCC1=CC=C2C=CC=CC2=C1 |
| C12H9N | |
| MFCD00004122 | |
| 2-naphthylacetonitrile, 2-naphthalen-2-yl acetonitrile, 2-naphthaleneacetonitrile, 2-naphthyl acetonitrile, naphthalene-2-acetonitrile, 2-2-naphthyl acetonitrile, .beta.-naphthyleneacetonitrile, 2-naphtylacetonitrile, 2-napthylacetonitrile, 2-naphthylmethylcyanide | |
| 2-naphthalen-2-ylacetonitrile |
Safety and Handling
GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 231-352-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only