missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthylacetonitrile, 97%

Catalog No. AAL0938406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0938406 5 g
AAL0938414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0938406 Supplier Thermo Scientific Chemicals Supplier No. L0938406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7498-57-9 | C12H9N | 167.21 g/mol

2-Naphthylacetonitrile was used as starting reagent in the synthesis of 2-(1-cyano-1-(2-naphthyl))methylidene-3-phenyl-thiazolidine-4,5-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthylacetonitrile was used as starting reagent in the synthesis of 2-(1-cyano-1-(2-naphthyl))methylidene-3-phenyl-thiazolidine-4,5-dione.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 7498-57-9
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.21
MDL Number MFCD00004122
InChI Key LPCWDVLDJVZIHA-UHFFFAOYSA-N
Synonym 2-naphthylacetonitrile, 2-naphthalen-2-yl acetonitrile, 2-naphthaleneacetonitrile, 2-naphthyl acetonitrile, naphthalene-2-acetonitrile, 2-2-naphthyl acetonitrile, .beta.-naphthyleneacetonitrile, 2-naphtylacetonitrile, 2-napthylacetonitrile, 2-naphthylmethylcyanide
PubChem CID 82008
IUPAC Name 2-naphthalen-2-ylacetonitrile
SMILES N#CCC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 82°C to 84°C
Density 1.092
Boiling Point 303°C
Quantity 5 g
UN Number UN3439
Beilstein 1862879
Solubility Information Insoluble in water.
Formula Weight 167.21
Percent Purity 97%
Chemical Name or Material 2-Naphthylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 231-352-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only