missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthylacetic acid, 99%

Catalog No. AAA1352818
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1352818 50 g
AAA1352806 5 g
AAA1352809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1352818 Supplier Thermo Scientific Chemicals Supplier No. A1352818

missing translation for 'validMainframeMsg'

CAS: 581-96-4 | C12H10O2 | 186.21 g/mol

2-Naphthylacetic acid is a synthetic plant hormone in the auxin family and is an ingredient in many commercial plant rooting horticultural products, it is a rooting agent and used for the vegetative propagation of plants from stem and leaf cutting. It is also used for plant tissue culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthylacetic acid is a synthetic plant hormone in the auxin family and is an ingredient in many commercial plant rooting horticultural products, it is a rooting agent and used for the vegetative propagation of plants from stem and leaf cutting. It is also used for plant tissue culture.

Solubility
Soluble in acetone (50 mg/ml), and methanol. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 581-96-4
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004126
InChI Key VIBOGIYPPWLDTI-UHFFFAOYSA-N
Synonym 2-naphthylacetic acid, 2-naphthaleneacetic acid, 2-naphthalen-2-yl acetic acid, betoxan, 2-2-naphthyl acetic acid, beta-naphthylacetic acid, beta-naphthaleneacetic acid, beta-naphthaleneacetate, naphthalen-2-ylacetic acid, .beta.-naphthaleneacetic acid
PubChem CID 11393
ChEBI CHEBI:37837
IUPAC Name 2-naphthalen-2-ylacetic acid
SMILES OC(=O)CC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 141°C to 143°C
Quantity 50 g
Beilstein 973396
Solubility Information Soluble in acetone (50mg/ml),and methanol. Insoluble in water.
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 2-Naphthylacetic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 209-475-0
RTECSNumber QJ0876000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only