missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Naphthaleneacetic Acid 98.0+%, TCI America™
$76.43 - $255.38
Chemical Identifiers
| CAS | 581-96-4 |
|---|---|
| Molecular Formula | C12H10O2 |
| Molecular Weight (g/mol) | 186.21 |
| MDL Number | MFCD00004126 |
| InChI Key | VIBOGIYPPWLDTI-UHFFFAOYSA-N |
| Synonym | 2-naphthylacetic acid, 2-naphthaleneacetic acid, 2-naphthalen-2-yl acetic acid, betoxan, 2-2-naphthyl acetic acid, beta-naphthylacetic acid, beta-naphthaleneacetic acid, beta-naphthaleneacetate, naphthalen-2-ylacetic acid, .beta.-naphthaleneacetic acid |
| PubChem CID | 11393 |
| ChEBI | CHEBI:37837 |
| IUPAC Name | 2-(naphthalen-2-yl)acetic acid |
| SMILES | OC(=O)CC1=CC=C2C=CC=CC2=C1 |
Chemical Identifiers
| 581-96-4 | |
| 186.21 | |
| VIBOGIYPPWLDTI-UHFFFAOYSA-N | |
| 11393 | |
| 2-(naphthalen-2-yl)acetic acid |
| C12H10O2 | |
| MFCD00004126 | |
| 2-naphthylacetic acid, 2-naphthaleneacetic acid, 2-naphthalen-2-yl acetic acid, betoxan, 2-2-naphthyl acetic acid, beta-naphthylacetic acid, beta-naphthaleneacetic acid, beta-naphthaleneacetate, naphthalen-2-ylacetic acid, .beta.-naphthaleneacetic acid | |
| CHEBI:37837 | |
| OC(=O)CC1=CC=C2C=CC=CC2=C1 |
Specifications
| 581-96-4 | |
| White-Yellow | |
| MFCD00004126 | |
| 2-naphthylacetic acid, 2-naphthaleneacetic acid, 2-naphthalen-2-yl acetic acid, betoxan, 2-2-naphthyl acetic acid, beta-naphthylacetic acid, beta-naphthaleneacetic acid, beta-naphthaleneacetate, naphthalen-2-ylacetic acid, .beta.-naphthaleneacetic acid | |
| OC(=O)CC1=CC=C2C=CC=CC2=C1 | |
| 186.21 | |
| CHEBI:37837 | |
| ≥98.0% (GC,T) | |
| 2-Naphthaleneacetic Acid |
| 142°C | |
| C12H10O2 | |
| 5 g | |
| VIBOGIYPPWLDTI-UHFFFAOYSA-N | |
| 2-(naphthalen-2-yl)acetic acid | |
| 11393 | |
| 186.21 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : QJ0876000
TSCA : No