missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthaleneacetic Acid 98.0+%, TCI America™

Catalog No. N03525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03525G 5 g
N035225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03525G Supplier TCI America Supplier No. N03525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 581-96-4
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004126
InChI Key VIBOGIYPPWLDTI-UHFFFAOYSA-N
Synonym 2-naphthylacetic acid, 2-naphthaleneacetic acid, 2-naphthalen-2-yl acetic acid, betoxan, 2-2-naphthyl acetic acid, beta-naphthylacetic acid, beta-naphthaleneacetic acid, beta-naphthaleneacetate, naphthalen-2-ylacetic acid, .beta.-naphthaleneacetic acid
PubChem CID 11393
ChEBI CHEBI:37837
IUPAC Name 2-(naphthalen-2-yl)acetic acid
SMILES OC(=O)CC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthaleneacetic Acid
RTECSNumber QJ0876000
TSCA No