missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Naphthaldehyde 98.0+%, TCI America™
$63.38 - $244.19
Chemical Identifiers
| CAS | 66-99-9 |
|---|---|
| Molecular Formula | C11H8O |
| Molecular Weight (g/mol) | 156.184 |
| MDL Number | MFCD00004094 |
| InChI Key | PJKVFARRVXDXAD-UHFFFAOYSA-N |
| Synonym | 2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene |
| PubChem CID | 6201 |
| ChEBI | CHEBI:52368 |
| IUPAC Name | naphthalene-2-carbaldehyde |
| SMILES | C1=CC=C2C=C(C=CC2=C1)C=O |
Chemical Identifiers
| 66-99-9 | |
| 156.184 | |
| PJKVFARRVXDXAD-UHFFFAOYSA-N | |
| 6201 | |
| naphthalene-2-carbaldehyde |
| C11H8O | |
| MFCD00004094 | |
| 2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene | |
| CHEBI:52368 | |
| C1=CC=C2C=C(C=CC2=C1)C=O |
Specifications
| 66-99-9 | |
| White-Yellow | |
| C11H8O | |
| 5 g | |
| PJKVFARRVXDXAD-UHFFFAOYSA-N | |
| naphthalene-2-carbaldehyde | |
| 6201 | |
| 156.18 | |
| Crystalline Powder |
| 60°C | |
| 147°C | |
| MFCD00004094 | |
| 2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene | |
| C1=CC=C2C=C(C=CC2=C1)C=O | |
| 156.184 | |
| CHEBI:52368 | |
| ≥98.0% (GC) | |
| 2-Naphthaldehyde |
Safety and Handling
RTECSNumber : QJ0190010
TSCA : No