missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthaldehyde, 98%

Catalog No. AAB2515706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2515706 5 g
AAB2515714 25 g
AAB2515722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2515706 Supplier Thermo Scientific Chemicals Supplier No. B2515706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 66-99-9 | C11H8O | 156.184 g/mol

2-Naphthaldehyde aids in stereo-selective formation of bicyclo 2-siloxy-2-alkoxyoxetanes. It is also used as catalytic agent and petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthaldehyde aids in stereo-selective formation of bicyclo 2-siloxy-2-alkoxyoxetanes. It is also used as catalytic agent and petrochemical additive.

Solubility
Soluble in hot water.

Notes
Store at -20°C. Air Sensitive. Store away from string bases and oxidizing agents.

Chemical Identifiers

CAS 66-99-9
Molecular Formula C11H8O
Molecular Weight (g/mol) 156.184
MDL Number MFCD00004094
InChI Key PJKVFARRVXDXAD-UHFFFAOYSA-N
Synonym 2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene
PubChem CID 6201
ChEBI CHEBI:52368
IUPAC Name naphthalene-2-carbaldehyde
SMILES C1=CC=C2C=C(C=CC2=C1)C=O

Specifications

Melting Point 59°C to 62°C
Boiling Point 160°C (19 mmHg)
Quantity 5 g
Beilstein 507750
Sensitivity Air Sensitive
Solubility Information Soluble in hot water.
Formula Weight 156.18
Percent Purity 98%
Chemical Name or Material 2-Naphthaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 200-640-2
RTECSNumber QJ0190010
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only