missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propyl-1-pentanol, 97%

Catalog No. AAH5795403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5795403 1 g
AAH5795406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5795403 Supplier Thermo Scientific Chemicals Supplier No. H5795403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58175-57-8 | C8H18O | 130.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58175-57-8
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00010260
InChI Key LASHFHLFDRTERB-UHFFFAOYSA-N
Synonym 2-propyl-1-pentanol, 1-pentanol, 2-propyl, 2-propylpentanol, 2-n-propyl-1-pentanol, acmc-1azt3, 2-propyl-1-hydroxypentane
PubChem CID 123543
IUPAC Name 2-propylpentan-1-ol
SMILES CCCC(CCC)CO

Specifications

Color Colorless to Yellow
Density 0.83 g/mL
Boiling Point 80°C to 82°C (12 mmHg)
Flash Point 72°C (162°F)
Refractive Index 1.43
Linear Formula (CH3CH2CH2)2CHCH2OH
Quantity 1 g
Formula Weight 130.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-n-Propyl-1-pentanol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
RTECSNumber SA7660000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only