missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propyl-1-heptanol, 98%

Catalog No. AAH5034003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5034003 1 g
AAH5034006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5034003 Supplier Thermo Scientific Chemicals Supplier No. H5034003

CAS: 10042-59-8 | C10H22O | 158.285 g/mol

2-n-Propyl-1-heptanol is used as a starting material for the production of plasticizers for PVC, e.g. Palatinol™ 10-P.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Propyl-1-heptanol is used as a starting material for the production of plasticizers for PVC, e.g. Palatinol™ 10-P.

Solubility
Insoluble in water. Miscible with most common organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 10042-59-8
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00046768
InChI Key YLQLIQIAXYRMDL-UHFFFAOYSA-N
Synonym 2-propyl-1-heptanol, 1-heptanol, 2-propyl, 2-propylheptanol, 2-n-propyl-1-heptanol, 2-propylheptyl alcohol, 2-propyl heptanol, 2-propyl-heptan-1-ol, acmc-20ann4, 2-propyl-1-heptanol #, dsstox_cid_9302
PubChem CID 24847
IUPAC Name 2-propylheptan-1-ol
SMILES CCCCCC(CCC)CO

Specifications

Quantity 1 g
Solubility Information Insoluble in water. Miscible with most common organic solvents.
Formula Weight 158.28
Percent Purity 98%
Chemical Name or Material 2-n-Propyl-1-heptanol
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 233-126-1
RTECSNumber MJ4800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only