missing translation for 'onlineSavingsMsg'
Learn More

2-n-Octylthiophene 98.0+%, TCI America™

Catalog No. O03765G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03765G Supplier TCI America Supplier No. O03765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 880-36-4
Molecular Formula C12H20S
Molecular Weight (g/mol) 196.352
MDL Number MFCD00041015
InChI Key GIFWAJGKWIDXMY-UHFFFAOYSA-N
Synonym 2-n-octylthiophene, thiophene, 2-octyl, octylthiophene, acmc-209qqw
PubChem CID 70153
IUPAC Name 2-octylthiophene
SMILES CCCCCCCCC1=CC=CS1

Specifications

Color Yellow
Boiling Point 125°C
Quantity 5 g
Formula Weight 196.35
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-n-Octylthiophene
TSCA No
Recommended Storage Refrigerator