missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-n-Octylbenzotriazole 98.0+%, TCI America™
$229.29 - $680.38
Chemical Identifiers
| CAS | 112642-69-0 |
|---|---|
| Molecular Formula | C14H21N3 |
| Molecular Weight (g/mol) | 231.343 |
| InChI Key | WESNTVKWORZVQQ-UHFFFAOYSA-N |
| PubChem CID | 4371641 |
| IUPAC Name | 2-octylbenzotriazole |
| SMILES | CCCCCCCCN1N=C2C=CC=CC2=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
O0430200MG
|
TCI America
O0430200MG |
200 mg |
Each for $229.29
|
|
|||||
|
O04301G
|
TCI America
O04301G |
1 g |
Each for $680.38
|
|
|||||
Chemical Identifiers
| 112642-69-0 | |
| 231.343 | |
| 4371641 | |
| CCCCCCCCN1N=C2C=CC=CC2=N1 |
| C14H21N3 | |
| WESNTVKWORZVQQ-UHFFFAOYSA-N | |
| 2-octylbenzotriazole |
Specifications
| 112642-69-0 | |
| 110°C | |
| 200 mg | |
| CCCCCCCCN1N=C2C=CC=CC2=N1 | |
| 231.343 | |
| 231.34 | |
| Liquid |
| Yellow | |
| C14H21N3 | |
| WESNTVKWORZVQQ-UHFFFAOYSA-N | |
| 2-octylbenzotriazole | |
| 4371641 | |
| ≥98.0% (GC) | |
| 2-n-Octylbenzotriazole |
Safety and Handling
TSCA : No