missing translation for 'onlineSavingsMsg'
Learn More

2-n-Octylbenzotriazole 98.0+%, TCI America™

Catalog No. O0430200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0430200MG 200 mg
O04301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0430200MG Supplier TCI America Supplier No. O0430200MG

Chemical Identifiers

CAS 112642-69-0
Molecular Formula C14H21N3
Molecular Weight (g/mol) 231.343
InChI Key WESNTVKWORZVQQ-UHFFFAOYSA-N
PubChem CID 4371641
IUPAC Name 2-octylbenzotriazole
SMILES CCCCCCCCN1N=C2C=CC=CC2=N1

Specifications

Color Yellow
Boiling Point 110°C
Quantity 200 mg
Formula Weight 231.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-n-Octylbenzotriazole